Stampa…

4,6-Dibromopyrimidine

Fornitore: Apollo Scientific

OR310203-1G OR310203-500MG
APOSOR310203-1GEA 611 EUR
APOSOR310203-1G APOSOR310203-500MG
4,6-Dibromopyrimidine
4,6-Dibromopyrimidine
Formula: C₄H₂Br₂N₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00233946
CAS Number: 36847-10-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR