4'-Demethylepipodophyllotoxin
Fornitore: Apollo Scientific
Synonyms:
(5R,5aR,8aR,9S)-9-Hydroxy-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one
OR3101T-5G
OR3101T-1G
OR3101T-10G
APOSOR3101T-5GEA
118
EUR
APOSOR3101T-5G
APOSOR3101T-1G
APOSOR3101T-10G
4'-Demethylepipodophyllotoxin
4'-Demethylepipodophyllotoxin
A potent inhibitor of microtubule assembly.
Formula:
C₂₁H₂₀O₈ Peso molecolare: 400,38 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00189421 CAS Number: 6559-91-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...