Stampa…

3-Bromoveratrole

Fornitore: Apollo Scientific

Synonyms: 1-Bromo-2,3-dimethoxybenzene

OR310088-1G OR310088-500MG
APOSOR310088-1GEA 364 EUR
APOSOR310088-1G APOSOR310088-500MG
3-Bromoveratrole
3-Bromoveratrole
Formula: C₈H₉BrO₂
Peso molecolare: 217,06 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00185638
CAS Number: 5424-43-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR