Stampa…

2,4-Dibenzyloxypyrimidine

Fornitore: Apollo Scientific

Synonyms: 2,4-Bis(benzyloxy)pyrimidine

OR310085-1G OR310085-500MG
APOSOR310085-1GEA 364 EUR
APOSOR310085-1G APOSOR310085-500MG
2,4-Dibenzyloxypyrimidine
2,4-Dibenzyloxypyrimidine
Formula: C₁₈H₁₆N₂O₂
Peso molecolare: 292,34 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06798236
CAS Number: 7306-79-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR