Stampa…

2-Amino-5-iodo-4-methylpyridine

Fornitore: Apollo Scientific

Synonyms: 5-Iodo-4-methyl-pyridin-2-ylamine, 5-Iodo-4-methylpyridin-2-ylamine

OR310075-1G OR310075-500MG
APOSOR310075-1GEA 364 EUR
APOSOR310075-1G APOSOR310075-500MG
2-Amino-5-iodo-4-methylpyridine
2-Amino-5-iodo-4-methylpyridine
Formula: C₆H₇IN₂
Peso molecolare: 234,04 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06637727
CAS Number: 356561-08-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR