3-Chloroquinoxaline-2-carboxylic acid
Fornitore: Apollo Scientific
OR310057-1G
OR310057-250MG
APOSOR310057-1GEA
183
EUR
APOSOR310057-1G
APOSOR310057-250MG
3-Chloroquinoxaline-2-carboxylic acid
3-Chloroquinoxaline-2-carboxylic acid
Formula:
C9H5ClN2O2 Peso molecolare: 208,6000 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD09864191 CAS Number: 20254-76-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...