Ethyl-6-chloro-4-hydroxyquinoline-3-carboxylate
Fornitore: Apollo Scientific
Synonyms:
6-Chloro-4-hydroxyquinoline-3-carboxylic acid ethyl ester, Ethyl 6-chloro-4-hydroxyquinoline-3-carboxylate
OR309457-1G
OR309457-5G
APOSOR309457-1GEA
85.8
EUR
APOSOR309457-1G
APOSOR309457-5G
Ethyl-6-chloro-4-hydroxyquinoline-3-carboxylate
Ethyl-6-chloro-4-hydroxyquinoline-3-carboxylate
Formula:
C₁₂H₁₀ClNO₃ Peso molecolare: 251,6657 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00173342 CAS Number: 70271-77-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...