1-Boc-DL-nipecotic acid ethyl ester
Fornitore: Apollo Scientific
Synonyms:
Ethyl 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate, Ethyl 1-Boc-DL-nipecotate, Ethyl 1-Boc-3-piperidinecarboxylate
OR309162-1G
OR309162-5G
APOSOR309162-1GEA
109
EUR
APOSOR309162-1G
APOSOR309162-5G
1-Boc-DL-nipecotic acid ethyl ester
1-Boc-DL-nipecotic acid ethyl ester
Formula:
C₁₃H₂₃NO₄ Peso molecolare: 257,33 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD04116274 CAS Number: 130250-54-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...