6-Bromo-4-chloro-2-propylquinoline
Fornitore: Apollo Scientific
OR307870-1G
OR307870-5G
APOSOR307870-1GEA
259
EUR
APOSOR307870-1G
APOSOR307870-5G
6-Bromo-4-chloro-2-propylquinoline
6-Bromo-4-chloro-2-propylquinoline
Formula:
C₁₂H₁₁BrClN Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09260431 CAS Number: 930570-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...