2-Methyl-4,7,8-trichloroquinoline
Fornitore: Apollo Scientific
Synonyms:
4,7,8-Trichloro-2-methylquinoline
OR307850-1G
OR307850-5G
APOSOR307850-1GEA
219
EUR
APOSOR307850-1G
APOSOR307850-5G
2-Methyl-4,7,8-trichloroquinoline
2-Methyl-4,7,8-trichloroquinoline
Formula:
C₁₀H₆Cl₃N Peso molecolare: 246,520 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00272309 CAS Number: 108097-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...