2-Methyl-4,6,8-trichloroquinoline
Fornitore: Apollo Scientific
OR307849-1G
OR307849-5G
APOSOR307849-1GEA
219
EUR
APOSOR307849-1G
APOSOR307849-5G
2-Methyl-4,6,8-trichloroquinoline
2-Methyl-4,6,8-trichloroquinoline
Formula:
C₁₀H₆Cl₃N Temperatura di stoccaggio: Ambiente |
CAS Number:
1204-14-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...