Stampa…

4-(Pyrrolidin-1-ylmethyl)benzoic acid hydrochloride

Fornitore: Apollo Scientific

OR303937-1G OR303937-500MG
APOSOR303937-1GEA 187 EUR
APOSOR303937-1G APOSOR303937-500MG
4-(Pyrrolidin-1-ylmethyl)benzoic acid hydrochloride
4-(Pyrrolidin-1-ylmethyl)benzoic acid hydrochloride
Formula: C₁₂H₁₆ClNO₂
Punto di fusione: 236…240 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD15201458
CAS Number: 159691-25-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR