Stampa…

2-(1-Lamda-{2}-piperidin-4-yl)pyridin-4-yl dihydrochloride

Fornitore: Apollo Scientific

Synonyms: 2-(Piperidin-4-yl)pyridine dihydrochloride

OR303856-1G OR303856-250MG
APOSOR303856-1GEA 448 EUR
APOSOR303856-1G APOSOR303856-250MG
2-(1-Lamda-{2}-piperidin-4-yl)pyridin-4-yl dihydrochloride
2-(1-Lamda-{2}-piperidin-4-yl)pyridin-4-yl dihydrochloride
Formula: C₁₀H₁₆Cl₂N₂
Peso molecolare: 235,16 g/mol
Punto di fusione: 258…261 °C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD14583122
CAS Number: 143924-45-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR