1-Naphthyl boronic acid pinacol ester
Fornitore: Apollo Scientific
Synonyms:
Naphthalene-1-boronic acid Pinacol ester, 1-Naphthalenboronic acid, pinacol ester, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene, 4,4,5,5-Tetramethyl-2-(1-naphthyl)-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-(naphthalen-1-yl)-1,3,2-dioxaborolane
Formula:
C₁₆H₁₉BO₂ Peso molecolare: 254,1319 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD05663869 CAS Number: 68716-52-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...