Stampa…

4-(Methylthio)phenylboronic acid pinacol ester

Fornitore: Apollo Scientific

Synonyms: 4,4,5,5-Tetramethyl-2-(4-(methylthio)phenyl)-1,3,2-dioxaborolane

OR303555-1G OR303555-5G
APOSOR303555-1GEA 78.6 EUR
APOSOR303555-1G APOSOR303555-5G
4-(Methylthio)phenylboronic acid pinacol ester
4-(Methylthio)phenylboronic acid pinacol ester
Formula: C₁₃H₁₉BO₂S
Peso molecolare: 250,1648 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD05155222
CAS Number: 190788-58-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR