Stampa…

3-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester 98%

Fornitore: Apollo Scientific
Attenzione

Synonyms: 3-{[4-(tert-Butoxycarbonyl)piperazin-1-yl]methyl}benzeneboronic acid, pinacol ester

OR303489-1G OR303489-5G
APOSOR303489-1GEA 81.5 EUR
APOSOR303489-1G APOSOR303489-5G
3-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester 98%
3-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester
Formula: C₂₂H₃₅BN₂O₄
Peso molecolare: 402,34 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD16294542
CAS Number: 883738-19-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR