Stampa…

1-(5-Amino-1-methyl-1H-pyrazol-4-yl)ethan-1-one

Fornitore: Apollo Scientific

OR303053-1G OR303053-500MG
APOSOR303053-1GEA 415 EUR
APOSOR303053-1G APOSOR303053-500MG
1-(5-Amino-1-methyl-1H-pyrazol-4-yl)ethan-1-one
1-(5-Amino-1-methyl-1H-pyrazol-4-yl)ethan-1-one
Formula: C₆H₉N₃O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD19213343
CAS Number: 856860-17-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR