O-[(Benzyloxy)methyl]hydroxylamine
Fornitore: Apollo Scientific
|
Attenzione
|
Formula:
C₈H₁₁NO₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD11111598 CAS Number: 895588-70-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...