5-Chloro-1-methyl-4-nitro-1H-pyrazole 97%
Fornitore: Apollo Scientific
|
Attenzione
|
OR302127-1G
OR302127-250MG
APOSOR302127-1GEA
115
EUR
APOSOR302127-1G
APOSOR302127-250MG
5-Chloro-1-methyl-4-nitro-1H-pyrazole 97%
5-Chloro-1-methyl-4-nitro-1H-pyrazole
Formula:
C₄H₄ClN₃O₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD21607270 CAS Number: 42098-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...