Stampa…

Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate 95%

Fornitore: Apollo Scientific

Synonyms: Ethyl 1-benzyl-3-oxopiperidine-4-carboxylate

OR301117-5G OR301117-25G
APOSOR301117-5GEA 85.8 EUR
APOSOR301117-5G APOSOR301117-25G
Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate 95%
Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate
Formula: C₁₅H₁₉NO₃
Peso molecolare: 261,32 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00044512
CAS Number: 39514-19-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR