Stampa…

Methyl benzimidate hydrochloride

Fornitore: Apollo Scientific

Synonyms: Benzimidic Acid Methyl Ester Hydrochloride

OR300905-1G OR300905-5G
APOSOR300905-1GEA 65.5 EUR
APOSOR300905-1G APOSOR300905-5G
Methyl benzimidate hydrochloride
Methyl benzimidate hydrochloride
Formula: C₈H₉NO
Peso molecolare: 171,62 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00012575
CAS Number: 5873-90-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR