Stampa…

(Phenylthio)acetonitrile

Fornitore: Apollo Scientific

Synonyms: 2-(Phenylthio)acetonitrile

OR30082-10G OR30082-1G OR30082-5G
APOSOR30082-10GEA 87.3 EUR
APOSOR30082-10G APOSOR30082-1G APOSOR30082-5G
(Phenylthio)acetonitrile
(Phenylthio)acetonitrile
Formula: C₈H₇NS
Peso molecolare: 149,22 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00019831
CAS Number: 5219-61-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR