Piperidine-1-carboxamidinium acetate
Fornitore: Apollo Scientific
Formula:
C₈H₁₇N₃O₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09864982 CAS Number: 92658-58-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...