Stampa…

5-Amino-2-chlorobenzothiazole 98%

Fornitore: Apollo Scientific

OR300771-1G OR300771-250MG OR300771-5G
APOSOR300771-1GEA 154 EUR
APOSOR300771-1G APOSOR300771-250MG APOSOR300771-5G
5-Amino-2-chlorobenzothiazole 98%
5-Amino-2-chlorobenzothiazole
Formula: C₇H₅ClN₂S
Peso molecolare: 184,6460 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11040282
CAS Number: 80945-82-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR