Stampa…

(4-Iodophenoxy)acetic acid

Fornitore: Apollo Scientific

OR300666-25G OR300666-5G
APOSOR300666-25GEA 31.6 EUR
APOSOR300666-25G APOSOR300666-5G
(4-Iodophenoxy)acetic acid
(4-Iodophenoxy)acetic acid
Formula: (C₆H₄I)OCH₂COOH
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00014359
CAS Number: 1878-94-0

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR