2-Amino-3-chloro-5-iodopyrazine
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
3-Chloro-5-iodopyrazin-2-amine
Formula:
C₄H₃ClIN₃ Peso molecolare: 255.45 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD14706048 CAS Number: 1252597-70-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...