Stampa…

1-Amino-4-bromoisoquinoline 97%

Fornitore: Apollo Scientific

Synonyms: 4-Bromoisoquinolin-1-amine

OR300056-1G OR300056-25G OR300056-5G
APOSOR300056-1GEA 75.7 EUR
APOSOR300056-1G APOSOR300056-25G APOSOR300056-5G
1-Amino-4-bromoisoquinoline 97%
1-Amino-4-bromoisoquinoline
Formula: C₉H₇BrN₂
Peso molecolare: 223,0693 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD05155261
CAS Number: 55270-27-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR