Stampa…

Trimethylsulphoxonium iodide 98%

Fornitore: Apollo Scientific
Attenzione

Synonyms: Trimethyloxosulphonium iodide

OR28785-1KG OR28785-250G
APOSOR28785-1KGEA 155 EUR
APOSOR28785-1KG APOSOR28785-250G
Trimethylsulphoxonium iodide 98%
Trimethylsulphoxonium iodide
Formula: C₃H₉IOS
Peso molecolare: 220,07 g/mol
Punto di fusione: 208…212 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011899
CAS Number: 1774-47-6
EINECS: 217-204-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR