Methyl-3,4-diaminobenzoate 98%
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
3,4-Diamino benzoate, 3,4-Diaminobenzoic acid methyl ester, Methyl 3,4-diaminobenzoate
OR26800-100G
OR26800-25G
OR26800-5G
APOSOR26800-100GEA
200
EUR
APOSOR26800-100G
APOSOR26800-25G
APOSOR26800-5G
Methyl-3,4-diaminobenzoate 98%
Methyl-3,4-diaminobenzoate
Formula:
C₈H₁₀N₂O₂ Peso molecolare: 166,18 g/mol Punto di fusione: 105…110 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00017098 CAS Number: 36692-49-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...