Stampa…

5-Chlorobenzo[b]thiophene-3-acetic acid

Fornitore: Apollo Scientific

OR26012-1G OR26012-250MG
APOSOR26012-1GEA 201 EUR
APOSOR26012-1G APOSOR26012-250MG
5-Chlorobenzo[b]thiophene-3-acetic acid
5-Chlorobenzo[b]thiophene-3-acetic acid
Formula: C₁₀H₇ClO₂S
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00052308
CAS Number: 17266-30-7

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR