5-Chlorosulphonyl-2-hydroxybenzoic acid
Fornitore: Apollo Scientific
|
Pericolo
|
OR24019-10G
OR24019-5G
APOSOR24019-10GEA
171
EUR
APOSOR24019-10G
APOSOR24019-5G
5-Chlorosulphonyl-2-hydroxybenzoic acid
5-Chlorosulphonyl-2-hydroxybenzoic acid
Formula:
C₇H₅ClO₅S Punto di fusione: 168…174 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00052974 CAS Number: 17243-13-9 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...