Stampa…

1-Benzyl-3-methylpiperidin-4-one 97%

Fornitore: Apollo Scientific
Pericolo

Synonyms: 1-Benzyl-3-methyl-4-piperidone

OR22901-100G OR22901-25G
APOSOR22901-100GEA 236 EUR
APOSOR22901-100G APOSOR22901-25G
1-Benzyl-3-methylpiperidin-4-one 97%
1-Benzyl-3-methylpiperidin-4-one
Formula: C₁₃H₁₇NO
Peso molecolare: 203,2800 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00044806
CAS Number: 34737-89-8
EINECS: 252-176-5
UN: 2735
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR