Stampa…

3,4,9,10-Perylenetetracarboxylic dianhydride 95%

Fornitore: Apollo Scientific

Synonyms: Anthra[2,1,9-def:6,5,10-d'e'f']diisochromene-1,3,8,10-tetraone, Pigment Red 224, PTCDA

OR22484-100G OR22484-1KG OR22484-500G
APOSOR22484-100GEA 118 EUR
APOSOR22484-100G APOSOR22484-1KG APOSOR22484-500G
3,4,9,10-Perylenetetracarboxylic dianhydride 95%
3,4,9,10-Perylenetetracarboxylic dianhydride
Formula: C₂₄H₈O₆
Peso molecolare: 392,32 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00006916
CAS Number: 128-69-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR