Stampa…

Dimethyl biphenyl-4,4'-dicarboxylate 99%

Fornitore: Apollo Scientific
Attenzione

Synonyms: Dimethyl [1,1'-biphenyl]-4,4'-dicarboxylate, 4,4'-Biphenyldicarboxylic Acid Dimethyl Ester, 4,4'-Bibenzoic Acid Dimethyl Ester, Dimethyl-4,4'-biphenyldicarboxylate

OR21762-25G OR21762-5G OR21762-100G
APOSOR21762-25GEA 72.7 EUR
APOSOR21762-25G APOSOR21762-5G APOSOR21762-100G
Dimethyl biphenyl-4,4'-dicarboxylate 99%
Dimethyl biphenyl-4,4'-dicarboxylate
Formula: C₁₆H₁₄O₄
Peso molecolare: 270,28 g/mol
Punto di fusione: 213…215 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00017201
CAS Number: 792-74-5
EINECS: 212-341-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR