Stampa…

5-Chlorosulphonyl-2-methoxybenzoic acid

Fornitore: Apollo Scientific
Pericolo

Synonyms: 5-(Chlorosulphonyl)-2-methoxybenzoic acid

OR2111-5G OR2111-1G OR2111-25G
APOSOR2111-5GEA 197 EUR
APOSOR2111-5G APOSOR2111-1G APOSOR2111-25G
5-Chlorosulphonyl-2-methoxybenzoic acid
5-Chlorosulphonyl-2-methoxybenzoic acid
Formula: C₈H₇ClO₅S
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00066912
CAS Number: 51904-91-7
UN: 3261
ADR: 8,II

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR