Cyclopropanecarboxamide 97%
Fornitore: Apollo Scientific
|
Attenzione
|
OR18286-25G
OR18286-100G
APOSOR18286-25GEA
52.4
EUR
APOSOR18286-25G
APOSOR18286-100G
Cyclopropanecarboxamide 97%
Cyclopropanecarboxamide
Formula:
C₄H₇NO Peso molecolare: 85,11 g/mol Punto di fusione: 120…122 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00013729 CAS Number: 6228-73-5 EINECS: 228-332-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...