3-Hydroxypicolinamide
Fornitore: Apollo Scientific
Formula:
C₆H₆N₂O₂ Punto di fusione: 193…197 °C Flash Pt: >110 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00006302 CAS Number: 933-90-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...