Stampa…

trans-4-Hydroxy-D-proline hydrochloride 97%

Fornitore: Apollo Scientific
Attenzione

Synonyms: (2R,4S)-4-Hydroxypyrrolidine-2-carboxylic acid hydrochloride, H-trans-D-Hyp-OH·HCl

OR17254-10G OR17254-1G OR17254-5G
APOSOR17254-10GEA 166 EUR
APOSOR17254-10G APOSOR17254-1G APOSOR17254-5G
trans-4-Hydroxy-D-proline hydrochloride 97%
trans-4-Hydroxy-D-proline hydrochloride
Formula: C₅H₁₀ClNO₃
Peso molecolare: 167,5908 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD08436165
CAS Number: 142347-81-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR