Stampa…

(-)-Di-p-toluoyl-L-tartaric acid 98%

Fornitore: Apollo Scientific
Attenzione

Synonyms: Di-p-toluoyl-L-tartaric acid, (-)-O,O'-Di-p-toluoyl-L-tartaric acid, (2R,3R)-(-)-Di-O-4-Toluoyl-L-Tartaric Acid

OR14901-1KG OR14901-500G
APOSOR14901-1KGEA 118 EUR
APOSOR14901-1KG APOSOR14901-500G
(-)-Di-p-toluoyl-L-tartaric acid 98%
(-)-Di-p-toluoyl-L-tartaric acid

Optical resolving agent.

Formula: C₂₀H₁₈O₈
Peso molecolare: 386,36 g/mol
Punto di fusione: 166…172 °C (dec.)
Densità: 1,138 g/cm³ (25 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00064440
CAS Number: 32634-66-5
EINECS: 251-131-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR