Stampa…

1-Phenylbut-3-yn-1-amine hydrochloride 95%

Fornitore: Apollo Scientific

Synonyms: (1-Aminobut-3-yn-1-yl)benzene hydrochloride

OR14851-1G OR14851-250MG
APOSOR14851-1GEA 619 EUR
APOSOR14851-1G APOSOR14851-250MG
1-Phenylbut-3-yn-1-amine hydrochloride 95%
1-Phenylbut-3-yn-1-amine hydrochloride
Formula: C₁₀H₁₂ClN
Peso molecolare: 181,67 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09865035
CAS Number: 1177093-09-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR