Stampa…

2-Aminothiazole-4-carboxylic acid ethyl ester hydrobromide

Fornitore: Apollo Scientific
Attenzione

Synonyms: Ethyl 2-amino-1,3-thiazole-4-carboxylate hydrobromide, Ethyl 2-aminothiazole-4-carboxylate hydrobromide

OR14763-1G OR14763-5G OR14763-10G
APOSOR14763-1GEA 14.7 EUR
APOSOR14763-1G APOSOR14763-5G APOSOR14763-10G
2-Aminothiazole-4-carboxylic acid ethyl ester hydrobromide
2-Aminothiazole-4-carboxylic acid ethyl ester hydrobromide
Formula: C₆H₉BrN₂O₂S
Peso molecolare: 253,12 g/mol
Punto di fusione: 175…177 °C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD06653384
CAS Number: 127942-30-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR