3,3-Diphenyl-L-alanine
Fornitore: Apollo Scientific
|
Pericolo
|
Synonyms:
H-Ala(3,3-DiPhenyl)-OH, (S)-2-Amino-3,3-diphenylpropanoic acid
OR14742-10G
OR14742-1G
OR14742-5G
APOSOR14742-10GEA
521
EUR
APOSOR14742-10G
APOSOR14742-1G
APOSOR14742-5G
3,3-Diphenyl-L-alanine
3,3-Diphenyl-L-alanine
Formula:
C₁₅H₁₅NO₂ Peso molecolare: 241,29 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD01631989 CAS Number: 149597-92-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...