Stampa…

3-Chloro-N-fmoc-L-phenylalanine

Fornitore: Apollo Scientific

Synonyms: Fmoc-Phe(3-Cl)-OH

OR14546-1G OR14546-5G
APOSOR14546-1GEA 59.7 EUR
APOSOR14546-1G APOSOR14546-5G
3-Chloro-N-fmoc-L-phenylalanine
3-Chloro-N-fmoc-L-phenylalanine
Formula: C₂₄H₂₀ClNO₄
Peso molecolare: 421,877 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00672548
CAS Number: 198560-44-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR