Stampa…

Veratrohydrazide

Fornitore: Apollo Scientific

Synonyms: 3,4-Dimethoxybenzohydrazide, 3,4-Dimethoxybenzene-1-carbohydrazide, 3,4-Dimethoxybenzoic hydrazide, 3,4-Dimethoxybenzoic acid hydrazide, 3,4-Dimethoxybenzhydrazide, 3,4-Dimethoxybenzenecarbohydrazide

OR14442-5G OR14442-25G OR14442-100G
APOSOR14442-5GEA 65.5 EUR
APOSOR14442-5G APOSOR14442-25G APOSOR14442-100G
Veratrohydrazide
Veratrohydrazide
Formula: C₉H₁₂N₂O₃
Peso molecolare: 196,21 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00017061
CAS Number: 41764-74-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR