3-(1,3-Benzodioxol-5-yl)-2-cyanoacrylic acid
Fornitore: Apollo Scientific
Formula:
C₁₁H₇NO₄ Punto di fusione: 170 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00759392 CAS Number: 49711-55-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...