Stampa…

N-Methyl-1-naphthylmethylamine hydrochloride

Fornitore: Apollo Scientific

Synonyms: Terbinafine Related Compound A, N-Methyl-1-(naphthalen-1-yl)methanamine hydrochloride, 1-(Methylaminomethyl)naphthalene Hydrochloride

OR13686-100G OR13686-25G
APOSOR13686-100GEA 147 EUR
APOSOR13686-100G APOSOR13686-25G
N-Methyl-1-naphthylmethylamine hydrochloride
N-Methyl-1-naphthylmethylamine hydrochloride
Formula: C₁₂H₁₃N
Peso molecolare: 207,70 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00012555
CAS Number: 65473-13-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR