Stampa…

4-Propoxycinnamic acid

Fornitore: Apollo Scientific

Synonyms: 3-(4-Propoxyphenyl)acrylic Acid

OR13168-10G OR13168-1G OR13168-5G
APOSOR13168-10GEA 112 EUR
APOSOR13168-10G APOSOR13168-1G APOSOR13168-5G
4-Propoxycinnamic acid
4-Propoxycinnamic acid
Formula: C₁₂H₁₄O₃
Peso molecolare: 206,24 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00995149
CAS Number: 69033-81-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR