Stampa…

4-(Piperidin-4-yloxy)pyridine dihydrochloride

Fornitore: Apollo Scientific
Attenzione

OR12807-1G OR12807-5G
APOSOR12807-1GEA 149 EUR
APOSOR12807-1G APOSOR12807-5G
4-(Piperidin-4-yloxy)pyridine dihydrochloride
4-(Piperidin-4-yloxy)pyridine dihydrochloride
Formula: C₁₀H₁₆Cl₂N₂O
Temperatura di stoccaggio: Ambiente
CAS Number: 308386-36-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR