Stampa…

Ethyl-2-cyanobutanoate 98%

Fornitore: Apollo Scientific
Attenzione

Synonyms: Ethyl 2-cyanobutanoate, Ethyl 2-cyanobutyrate

OR12466-10G OR12466-25G
APOSOR12466-10GEA 0 EUR
APOSOR12466-10G APOSOR12466-25G
Ethyl-2-cyanobutanoate 98%
Ethyl-2-cyanobutanoate
Formula: C₇H₁₁NO₂
Peso molecolare: 141,17 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD02258598
CAS Number: 1619-58-5
UN: 3276
ADR: 6.1,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR