Stampa…

2-Amino-N,N-dimethylacetamide hydrochloride 95%

Fornitore: Apollo Scientific
Attenzione

OR12015-1G OR12015-25G OR12015-5G
APOSOR12015-1GEA 65.5 EUR
APOSOR12015-1G APOSOR12015-25G APOSOR12015-5G
2-Amino-N,N-dimethylacetamide hydrochloride 95%
2-Amino-N,N-dimethylacetamide hydrochloride
Formula: C₄H₁₁ClN₂O
Peso molecolare: 138,5959 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD08060117
CAS Number: 72287-77-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR